3-Brom-5-chlor-2-hydroxy-4-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-5-chlor-2-hydroxy-4-picolin

PY-1237-5G PY-1237-1G
COBBPY-1237-5GEA 451 EUR
COBBPY-1237-5G COBBPY-1237-1G
3-Brom-5-chlor-2-hydroxy-4-methylpyridin
3-Bromo-5-chloro-2-hydroxy-4-methylpyridine
Formel: C₆H₅BrClNO
MDL: MFCD13195779
CAS-Nummer: 1199773-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR