3-Brom-5-chlor-2-methoxypyridin
Lieferant: COMBI-BLOCKS
PY-7789-1G
PY-7789-5G
COBBPY-7789-1GEA
38.2
EUR
COBBPY-7789-1G
COBBPY-7789-5G
3-Brom-5-chlor-2-methoxypyridin
3-Bromo-5-chloro-2-methoxypyridine
Formel:
C₆H₅BrClNO Molecular Weight: 222.47 g/mol |
MDL:
MFCD08059322 CAS-Nummer: 102830-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...