5-Brom-3-chlor-2-methoxypyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-3-chloro-5-bromopyridine

PY-7727-1G PY-7727-5G
COBBPY-7727-1GEA 28.7 EUR
COBBPY-7727-1G COBBPY-7727-5G
5-Brom-3-chlor-2-methoxypyridin
5-Brom-3-chlor-2-methoxypyridin
Formel: C₆H₅BrClNO
Molecular Weight: 222.47 g/mol
MDL: MFCD04972382
CAS-Nummer: 848366-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR