3-Brom-5-chlorpyridin

Lieferant: COMBI-BLOCKS

PY-7083-1G PY-7083-5G PY-7083-25G
COBBPY-7083-1GEA 26.2 EUR
COBBPY-7083-1G COBBPY-7083-5G COBBPY-7083-25G
3-Brom-5-chlorpyridin
3-Bromo-5-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
MDL: MFCD04114221
CAS-Nummer: 73583-39-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR