3-Brom-5-hydroxypyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
3-Brom-5-pyridinol
PY-7084-1G
PY-7084-25G
PY-7084-5G
COBBPY-7084-1GEA
25.5
EUR
COBBPY-7084-1G
COBBPY-7084-25G
COBBPY-7084-5G
3-Brom-5-hydroxypyridin
3-Brom-5-hydroxypyridin
Formel:
C₅H₄BrNO Molecular Weight: 174 g/mol |
MDL:
MFCD00661305 CAS-Nummer: 74115-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...