3-Brom-5-methoxypyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
5-Bromo-3-methoxypyridine
PY-7278-1G
PY-7278-25G
PY-7278-5G
COBBPY-7278-1GEA
0
EUR
COBBPY-7278-1G
COBBPY-7278-25G
COBBPY-7278-5G
3-Brom-5-methoxypyridin
3-Bromo-5-methoxypyridine
Formel:
C₆H₆BrNO Molecular Weight: 188.02 g/mol |
MDL:
MFCD00234169 CAS-Nummer: 50720-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...