Ethyl-3-brom-5-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-5-nitrobenzoesäureethylester

CA-4277-1G CA-4277-25G CA-4277-5G
COBBCA-4277-1GEA 75.9 EUR
COBBCA-4277-1G COBBCA-4277-25G COBBCA-4277-5G
Ethyl-3-brom-5-nitrobenzoat
Ethyl-3-brom-5-nitrobenzoat
Formel: C₉H₈BrNO₄
Molecular Weight: 274.07 g/mol
MDL: MFCD09258760
CAS-Nummer: 690260-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR