Methyl-3-brom-5-nitrobenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
3-Brom-5-nitrobenzoesäuremethylester
CA-4276-5G
CA-4276-25G
CA-4276-1G
COBBCA-4276-5GEA
103
EUR
COBBCA-4276-5G
COBBCA-4276-25G
COBBCA-4276-1G
Methyl-3-brom-5-nitrobenzoat
Methyl-3-brom-5-nitrobenzoat
Formel:
C₈H₆BrNO₄ Molecular Weight: 260,04 g/mol |
MDL:
MFCD09258759 CAS-Nummer: 6307-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...