3-Brom-6-chlor-2-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-chlor-6-picolin, 5-Brom-2-chlor-6-methylpyridin, 3-Brom-6-chlor-2-picolin

OT-0321-5G OT-0321-25G OT-0321-1G
COBBOT-0321-5GEA 50.9 EUR
COBBOT-0321-5G COBBOT-0321-25G COBBOT-0321-1G
3-Brom-6-chlor-2-methylpyridin
3-Bromo-6-chloro-2-methylpyridine
Formel: C₆H₅BrClN
Molecular Weight: 206,47 g/mol
MDL: MFCD07375117
CAS-Nummer: 132606-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR