5-Brom-2-chlor-4-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

PY-1163-1G PY-1163-5G PY-1163-250M
COBBPY-1163-1GEA 205 EUR
COBBPY-1163-1G COBBPY-1163-5G COBBPY-1163-250M
5-Brom-2-chlor-4-(trifluormethyl)pyridin
5-Brom-2-chlor-4-(trifluormethyl)pyridin
Formel: C₆H₂BrClF₃N
Molecular Weight: 260,44 g/mol
MDL: MFCD11100520
CAS-Nummer: 823221-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR