5-Brom-2-chlor-4-iodpyridin

Lieferant: COMBI-BLOCKS

PY-7284-1G PY-7284-5G
COBBPY-7284-1GEA 227 EUR
COBBPY-7284-1G COBBPY-7284-5G
5-Brom-2-chlor-4-iodpyridin
5-Brom-2-chlor-4-iodpyridin
Formel: C₅H₂BrClIN
Molecular Weight: 318.34 g/mol
MDL: MFCD09972189
CAS-Nummer: 401892-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR