5-Brom-2-chlor-4-methyl-3-nitropyridin
Lieferant: COMBI-BLOCKS
PY-7317-5G
PY-7317-1G
COBBPY-7317-5GEA
127
EUR
COBBPY-7317-5G
COBBPY-7317-1G
5-Brom-2-chlor-4-methyl-3-nitropyridin
5-Brom-2-chlor-4-methyl-3-nitropyridin
Formel:
C₆H₄BrClN₂O₂ Molecular Weight: 251,47 g/mol |
MDL:
MFCD06659505 CAS-Nummer: 884495-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...