5-Brom-2-chlor-4-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-chlor-4-picolin

OT-0318-5G OT-0318-25G OT-0318-1G
COBBOT-0318-5GEA 38.2 EUR
COBBOT-0318-5G COBBOT-0318-25G COBBOT-0318-1G
5-Brom-2-chlor-4-methylpyridin
5-Brom-2-chlor-4-methylpyridin
Formel: C₆H₅BrClN
Molecular Weight: 206.47 g/mol
MDL: MFCD03427663
CAS-Nummer: 778611-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR