5-Brom-2-chlor-3-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

PY-7980-1G PY-7980-5G
COBBPY-7980-1GEA 21.9 EUR
COBBPY-7980-1G COBBPY-7980-5G
5-Brom-2-chlor-3-(trifluormethyl)pyridin
5-Brom-2-chlor-3-(trifluormethyl)pyridin
Formel: C₆H₂BrClF₃N
Molecular Weight: 260,44 g/mol
MDL: MFCD11100983
CAS-Nummer: 211122-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR