5-Brom-2-chlor-3-fluorpyridin

Lieferant: COMBI-BLOCKS

PY-7782-1G PY-7782-25G PY-7782-5G
COBBPY-7782-1GEA 21.6 EUR
COBBPY-7782-1G COBBPY-7782-25G COBBPY-7782-5G
5-Brom-2-chlor-3-fluorpyridin
5-Brom-2-chlor-3-fluorpyridin
Formel: C₅H₂BrClFN
Molecular Weight: 210.43 g/mol
MDL: MFCD03411574
CAS-Nummer: 831203-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR