5-Brom-2-chlor-3-fluorpyridin

Lieferant: FLUOROCHEM

016231-5G 016231-1G
FLUO016231-5GEA 57.9 EUR
FLUO016231-5G FLUO016231-1G
5-Brom-2-chlor-3-fluorpyridin
5-Brom-2-chlor-3-fluorpyridin
Formel: C₅H₂BrClFN
MDL: MFCD03411574
CAS-Nummer: 831203-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR