5-Brom-2-chlor-3-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-chlor-3-picolin

PY-1119-5G PY-1119-1G PY-1119-25G
COBBPY-1119-5GEA 38 EUR
COBBPY-1119-5G COBBPY-1119-1G COBBPY-1119-25G
5-Brom-2-chlor-3-methylpyridin
5-Brom-2-chlor-3-methylpyridin
Formel: C₆H₅BrClN
Molecular Weight: 206.47 g/mol
MDL: MFCD03095093
CAS-Nummer: 29241-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR