5-Brom-2-chlor-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7174-100G PY-7174-5G PY-7174-25G
COBBPY-7174-100GEA 364 EUR
COBBPY-7174-100G COBBPY-7174-5G COBBPY-7174-25G
5-Brom-2-chlor-3-nitropyridin
5-Brom-2-chlor-3-nitropyridin
Formel: C₅H₂BrClN₂O₂
Molecular Weight: 237,44 g/mol
MDL: MFCD00222270
CAS-Nummer: 67443-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR