5-Brom-2-chlorpyridin

Lieferant: COMBI-BLOCKS
Achtung

OT-0324-100G OT-0324-25G OT-0324-5G
COBBOT-0324-100GEA 154 EUR
COBBOT-0324-100G COBBOT-0324-25G COBBOT-0324-5G
5-Brom-2-chlorpyridin
5-Brom-2-chlorpyridin
Formel: C₅H₃BrClN
Molecular Weight: 192,44 g/mol
Siedepunkt: 205 °C (1013 hPa)
Schmelzpunkt: 71 °C
MDL: MFCD01318951
CAS-Nummer: 53939-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR