5-Brom-2-fluor-3-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-fluor-3-picolin

SS-3015-1G SS-3015-25G SS-3015-5G
COBBSS-3015-1GEA 26.2 EUR
COBBSS-3015-1G COBBSS-3015-25G COBBSS-3015-5G
5-Brom-2-fluor-3-methylpyridin
5-Brom-2-fluor-3-methylpyridin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
MDL: MFCD03095091
CAS-Nummer: 29312-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR