5-Brom-2-fluorbenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

CA-4097-5G CA-4097-25G
COBBCA-4097-5GEA 26.2 EUR
COBBCA-4097-5G COBBCA-4097-25G
5-Brom-2-fluorbenzoesäure
5-Brom-2-fluorbenzoesäure
Formel: C₇H₄BrFO₂
Molecular Weight: 219,01 g/mol
MDL: MFCD00143423
CAS-Nummer: 146328-85-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR