5-Brom-3-methoxysalicylaldehyd
Lieferant: Thermo Scientific
Synonyme:
5-Brom-2-hydroxy-3-methoxybenzaldehyd
BTB03758.1
BTB03758EA
BTB03758EE
MAYBBTB03758.1EA
0
EUR
MAYBBTB03758.1
MAYBBTB03758EA
MAYBBTB03758EE
5-Brom-3-methoxysalicylaldehyd
5-Brom-3-methoxysalicylaldehyd
Formel:
C₈H₇BrO₃ Molecular Weight: 231.05 g/mol Siedepunkt: 314 Schmelzpunkt: 128 |
MDL:
MFCD00016593 CAS-Nummer: 5034-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...