5-Brom-3-methoxysalicylaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Brom-2-hydroxy-3-methoxybenzaldehyd
A10137.06
A10137.14
A10137.22
A10137.06EA
43.2
EUR
A10137.06
A10137.14
A10137.22
5-Brom-3-methoxysalicylaldehyd ≥98%
5-Brom-3-methoxysalicylaldehyd
Purity: 98%
CAS No.: 5034-74-2
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Melting Point: 127-131°
Beilstein Reference: 2328601
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol Siedepunkt: 314 °C (1013 hPa) Schmelzpunkt: 127…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016593 CAS-Nummer: 5034-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...