5-Brom-3-nitrosalicylaldehyd ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H26927.06 H26927.03
H26927.06EA 124 EUR
H26927.06 H26927.03
5-Brom-3-nitrosalicylaldehyd ≥97%
5-Brom-3-nitrosalicylaldehyd

Purity: 97% CAS No.: 16634-88-1 Molecular Formula: C7H4BrNO4 Formula Weight: 246.02 Melting Point: 125-128° MDL No.: MFCD00130110

Formel: C₇H₄BrNO₄
Schmelzpunkt: 125…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00130110
CAS-Nummer: 16634-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR