5-Brom-3-nitrosalicylaldehyd

Lieferant: CHEMPUR

Synonyme: 5-Brom-2-hydroxy-3-nitrobenzaldehyd

BTB14071.250 BTB14071.1
CHMPBTB14071.250EA 0 EUR
CHMPBTB14071.250 MAYBBTB14071.1
5-Brom-3-nitrosalicylaldehyd
5-Brom-3-nitrosalicylaldehyd
Formel: C₇H₄BrNO₄
Molecular Weight: 246,02 g/mol
MDL: MFCD00130110
CAS-Nummer: 16634-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR