5-Brom-2-iod-4-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-iod-4-picolin

PY-7157-5G PY-7157-1G
COBBPY-7157-5GEA 1150 EUR
COBBPY-7157-5G COBBPY-7157-1G
5-Brom-2-iod-4-methylpyridin
5-Brom-2-iod-4-methylpyridin
Formel: C₆H₅BrIN
Molecular Weight: 297.92 g/mol
MDL: MFCD09801049
CAS-Nummer: 941294-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR