5-Brom-2-iodpyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
CA-4303-5G
CA-4303-25G
CA-4303-100G
COBBCA-4303-5GEA
25.5
EUR
COBBCA-4303-5G
COBBCA-4303-25G
COBBCA-4303-100G
5-Brom-2-iodpyridin
5-Brom-2-iodpyridin
Formel:
C₅H₃BrIN Molecular Weight: 283,89 g/mol |
MDL:
MFCD00691506 CAS-Nummer: 223463-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...