5-Brom-2-methoxybenzolsulfonylchlorid

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-methoxybenzolsulfonsäurechlorid, 5-Brom-2-methoxybenzolsulfochlorid

OT-0196-25G OT-0196-5G
COBBOT-0196-25GEA 276 EUR
COBBOT-0196-25G COBBOT-0196-5G
5-Brom-2-methoxybenzolsulfonylchlorid
5-Brom-2-methoxybenzolsulfonylchlorid
Formel: C₇H₆BrClO₃S
Molecular Weight: 285.55 g/mol
Schmelzpunkt: 107…110 °C
MDL: MFCD00051768
CAS-Nummer: 23095-05-8
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR