5-Brom-2-methoxypyridin

Lieferant: COMBI-BLOCKS
Achtung

CA-4302-5G CA-4302-25G CA-4302-100G
COBBCA-4302-5GEA 25.5 EUR
COBBCA-4302-5G COBBCA-4302-25G COBBCA-4302-100G
5-Brom-2-methoxypyridin
5-Brom-2-methoxypyridin
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD01318952
CAS-Nummer: 13472-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR