5-Brom-3-nitro-2-picolin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Brom-2-methyl-3-nitropyridin
PY-1321-5G
PY-1321-1G
COBBPY-1321-5GEA
50.9
EUR
COBBPY-1321-5G
COBBPY-1321-1G
5-Brom-3-nitro-2-picolin
5-Brom-3-nitro-2-picolin
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217,02 g/mol |
MDL:
MFCD09031419 CAS-Nummer: 911434-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...