3-Brombenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 3-Bromphenylamidin Hydrochlorid, 3-Brombenzoesäureamidin Hydrochlorid

12-0055.500 12-0055.1
CHMP12-0055.500EA 0 EUR
CHMP12-0055.500 CHMP12-0055.1
3-Brombenzamidin Hydrochlorid
3-Brombenzamidin Hydrochlorid
Formel: C₇H₈BrClN₂
Molecular Weight: 235.51 g/mol
MDL: MFCD04114439
CAS-Nummer: 16796-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR