3-Brombenzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
3-Brombenzolsulfonsäureamid
OT-0155-5G
OT-0155-25G
OT-0155-1G
COBBOT-0155-5GEA
63
EUR
COBBOT-0155-5G
COBBOT-0155-25G
COBBOT-0155-1G
3-Brombenzolsulfonamid
3-Brombenzolsulfonamid
Formel:
C₆H₆BrNO₂S Molecular Weight: 236,09 g/mol |
MDL:
MFCD00084903 CAS-Nummer: 89599-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...