3-Brombenzolsulfonamid

Lieferant: Thermo Scientific

Synonyme: 3-Brombenzolsulfonsäureamid

TL00520DA TL00520EA
MAYBTL00520DAEA 0 EUR
MAYBTL00520DA MAYBTL00520EA
3-Brombenzolsulfonamid
3-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
MDL: MFCD00084903
CAS-Nummer: 89599-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR