3-Brombenzolsulfonamid ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Brombenzolsulfonsäureamid

B25681.03 B25681.06
B25681.03EA 31.4 EUR
B25681.03 B25681.06
3-Brombenzolsulfonamid ≥97%
3-Brombenzolsulfonamid

Purity: 97% CAS No.: 89599-01-9 Molecular Formula: C6H6BrNO2S Formula Weight: 236.09 Melting Point: 154-158° MDL No.: MFCD00084903

Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
Schmelzpunkt: 154…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084903
CAS-Nummer: 89599-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR