3-Chlor-2-fluorbenzoesäure
Lieferant: COMBI-BLOCKS
OS-7415-25G
OS-7415-100G
OS-7415-5G
COBBOS-7415-25GEA
101
EUR
COBBOS-7415-25G
COBBOS-7415-100G
COBBOS-7415-5G
3-Chlor-2-fluorbenzoesäure
3-Chloro-2-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Molecular Weight: 174,56 g/mol |
MDL:
MFCD00042506 CAS-Nummer: 161957-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...