3-Chlor-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chlor-2-fluorbenzolboronsäure, (3-Chlor-2-fluorphenyl)metaborsäure, Dihydroxy(3-chlor-2-fluorphenyl)boran

BB-2386-25G BB-2386-1G BB-2386-5G
COBBBB-2386-25GEA 247 EUR
COBBBB-2386-25G COBBBB-2386-1G COBBBB-2386-5G
3-Chlor-2-fluorphenylboronsäure
3-Chlor-2-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174.37 g/mol
MDL: MFCD05664224
CAS-Nummer: 352535-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR