3-Chlor-2-methoxypyridin

Lieferant: COMBI-BLOCKS

PY-7071-5G PY-7071-1G
COBBPY-7071-5GEA 143 EUR
COBBPY-7071-5G COBBPY-7071-1G
3-Chlor-2-methoxypyridin
3-Chloro-2-methoxypyridine
Formel: C₆H₆ClNO
Molecular Weight: 143.57 g/mol
MDL: MFCD03095227
CAS-Nummer: 13472-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR