3-Chlor-2-methylbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3-Chlor-2-methylbenzolsulfochlorid, 3-Chlor-2-methylbenzolsulfonsäurechlorid, 3-Chlor-2-methylbenzol-1-sulfonylchlorid
017439-10G
017439-25G
017439-100G
FLUO017439-10GEA
68.6
EUR
FLUO017439-10G
FLUO017439-25G
FLUO017439-100G
3-Chlor-2-methylbenzolsulfonylchlorid
3-Chlor-2-methylbenzolsulfonylchlorid
Formel:
C₇H₆Cl₂O₂S Molecular Weight: 225.09 Schmelzpunkt: 72 |
MDL:
MFCD00052198 CAS-Nummer: 80563-86-6 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...