3-Chlor-4-(trifluormethyl)benzoesäure 97%, JRD

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Chlor-α,α,α-trifluor-p-toluylsäure

H32038.03 H32038.06
H32038.03EA 35.2 EUR
H32038.03 H32038.06
3-Chlor-4-(trifluormethyl)benzoesäure 97%, JRD
3-Chloro-4-(trifluoromethyl)benzoic acid

Purity: 97% CAS No.: 115754-20-6 Molecular Formula: C8H4ClF3O2 Formula Weight: 224.57 Melting Point: 158-161° MDL No.: MFCD04972755

Formel: C₈H₄ClF₃O₂
Molecular Weight: 224,57 g/mol
Schmelzpunkt: 158…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972755
CAS-Nummer: 115754-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR