3-Chlor-4-fluorthiobenzamid 97
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Chlor-4-fluorthiobenzoesäureamid
L11132.03
L11132.MD
L11132.03EA
95
EUR
L11132.03
L11132.MD
3-Chlor-4-fluorthiobenzamid 97
3-Chlor-4-fluorthiobenzamid
Purity: 97%
CAS No.: 130560-97-3
Molecular Formula: C7H5ClFNS
Formula Weight: 189.64
Melting Point: 129-130°
Beilstein Reference: 7757675
UN No.: 2811
MDL No.: MFCD00042478
Formel:
C₇H₅ClFNS Molecular Weight: 189.64 g/mol Siedepunkt: 272 Schmelzpunkt: 129…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042478 CAS-Nummer: 130560-97-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...