3-Chlor-4-fluorthiobenzamid

Lieferant: CHEMPUR

Synonyme: 3-Chlor-4-fluorthiobenzoesäureamid

FL4468.POR 004468-1G
CHMPFL4468.POREA 0 EUR
CHMPFL4468.POR FLUO004468-1G
3-Chlor-4-fluorthiobenzamid
3-Chlor-4-fluorthiobenzamid
Formel: C₇H₅ClFNS
Molecular Weight: 189.64 g/mol
Siedepunkt: 272
Schmelzpunkt: 129…130 °C
MDL: MFCD00042478
CAS-Nummer: 130560-97-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR