3-Chlor-4-methylbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-p-toluolsulfochlorid, 3-Chlor-4-methylbenzolsulfonsäurechlorid, 3-Chlor-4-methylbenzolsulfochlorid, 3-Chlor-4-methylbenzol-1-sulfonylchlorid

013067-5G 013067-25G
FLUO013067-5GEA 40.1 EUR
FLUO013067-5G FLUO013067-25G
3-Chlor-4-methylbenzolsulfonylchlorid
3-Chlor-4-methylbenzolsulfonylchlorid
Formel: C₇H₆Cl₂O₂S
Molecular Weight: 225.09
MDL: MFCD00203987
CAS-Nummer: 42413-03-6
EINECS: 412-890-1
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR