3-Chlor-4-nitrobenzolsulfonylchlorid

Lieferant: CHEMPUR

Synonyme: 3-Chlor-4-nitrobenzolsulfonsäurechlorid, 3-Chlor-4-nitrobenzol-1-sulfonylchlorid, 3-Chlor-4-nitrobenzolsulfochlorid

FL13807.POR 013807-1G 013807-5G
CHMPFL13807.POREA 0 EUR
CHMPFL13807.POR FLUO013807-1G FLUO013807-5G
3-Chlor-4-nitrobenzolsulfonylchlorid
3-Chlor-4-nitrobenzolsulfonylchlorid
Formel: C₆H₃Cl₂NO₄S
Molecular Weight: 256.06 g/mol
MDL: MFCD04973767
CAS-Nummer: 64835-30-9
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR