Ethyl-3-chlor-5-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chlor-5-nitrobenzoesäureethylester

CA-4969-1G CA-4969-5G
COBBCA-4969-1GEA 149 EUR
COBBCA-4969-1G COBBCA-4969-5G
Ethyl-3-chlor-5-nitrobenzoat
Ethyl-3-chlor-5-nitrobenzoat
Formel: C₉H₈ClNO₄
Molecular Weight: 229.62 g/mol
MDL: MFCD12172957
CAS-Nummer: 1156940-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR