Methyl-3-chlor-5-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chlor-5-nitrobenzoesäuremethylester

CA-4335-5G CA-4335-1G
COBBCA-4335-5GEA 243 EUR
COBBCA-4335-5G COBBCA-4335-1G
Methyl-3-chlor-5-nitrobenzoat
Methyl-3-chlor-5-nitrobenzoat
Formel: C₈H₆ClNO₄
Molecular Weight: 215,59 g/mol
MDL: MFCD01529623
CAS-Nummer: 36138-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR