5-Chlor-2-(trifluormethyl)benzoesäure

Lieferant: FLUOROCHEM

Synonyme: 5-Chlor-α,α,α-trifluor-o-toluylsäure

005711-5G 005711-1G
FLUO005711-5GEA 0 EUR
FLUO005711-5G FLUO005711-1G
5-Chlor-2-(trifluormethyl)benzoesäure
5-Chlor-2-(trifluormethyl)benzoesäure
Formel: C₈H₄ClF₃O₂
Molecular Weight: 224,57 g/mol
MDL: MFCD01631353
CAS-Nummer: 654-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR