5-Chlor-2-fluor-4-iodpyridin

Lieferant: COMBI-BLOCKS

SS-3003-1G SS-3003-5G
COBBSS-3003-1GEA 63 EUR
COBBSS-3003-1G COBBSS-3003-5G
5-Chlor-2-fluor-4-iodpyridin
5-Chlor-2-fluor-4-iodpyridin
Formel: C₅H₂ClFIN
Molecular Weight: 257,43 g/mol
MDL: MFCD04112566
CAS-Nummer: 659731-48-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR