5-Chlor-2-methoxybenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 5-Chlor-2-methoxybenzolsulfochlorid, 5-Chlor-2-methoxybenzolsulfonsäurechlorid

TL00312.1 TL00312.10 TL00312.50
MAYBTL00312.1EA 0 EUR
MAYBTL00312.1 MAYBTL00312.10 MAYBTL00312.50
5-Chlor-2-methoxybenzolsulfonylchlorid
5-Chlor-2-methoxybenzolsulfonylchlorid
Formel: C₇H₆Cl₂O₃S
Molecular Weight: 241,09 g/mol
MDL: MFCD00052959
CAS-Nummer: 22952-32-5
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR