3-Chlor-6-methoxypyridazin
Lieferant: COMBI-BLOCKS
|
Achtung
|
HC-2807-1G
HC-2807-5G
HC-2807-25G
COBBHC-2807-1GEA
0
EUR
COBBHC-2807-1G
COBBHC-2807-5G
COBBHC-2807-25G
3-Chlor-6-methoxypyridazin
3-Chloro-6-methoxypyridazine
Formel:
C₅H₅ClN₂O Molecular Weight: 144,56 g/mol Siedepunkt: 174…175 °C (3 mmHg) Schmelzpunkt: 91 °C |
MDL:
MFCD00006467 CAS-Nummer: 1722-10-7 EINECS: 217-019-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...