3-Chlor-6-methylpyridazin

Lieferant: COMBI-BLOCKS

OS-7519-5G OS-7519-25G OS-7519-1G OS-7519-100G
COBBOS-7519-5GEA 96.8 EUR
COBBOS-7519-5G COBBOS-7519-25G COBBOS-7519-1G COBBOS-7519-100G
3-Chlor-6-methylpyridazin
3-Chloro-6-methylpyridazine
Formel: C₅H₅ClN₂
Molecular Weight: 128,56 g/mol
MDL: MFCD00052905
CAS-Nummer: 1121-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR